Agarose, LF™ for pulsed field gel electrophoresis (PFGE)
X174-250GEA 2469790 KRW
X174-250G X174-100G
Agarose, LF™ for pulsed field gel electrophoresis (PFGE)
Agarose
Agarose LF™ (Large Fragment, 1 kb - 40 kb) is optimised for pulsed-field gel electrophoresis (PFGE) applications. The exceptionally low EEO (electroendosmosis) and high gel strength of Agarose LF™ facilitates faster electrophoresis running times in low concentration gels. This agarose is best suited for resolving large (>20 kb) DNA fragments.
  • Optimised for pulsed field gel electrophoresis (PFGE)
  • Ideal for resolving large (>20 kb) DNA fragments
  • Nuclease and protease-free
끊는점: 1413 °C
녹는점: 60…90 °C
보관 온도: Ambient
MDL 번호: MFCD00081294
CAS 번호: 9012-36-6
EINECS: 232-731-8
UN: 0000
https://kr.vwr.com/stibo/web/std.lang.all/43/75/23914375.jpg
Order Now
사양 테스트 결과
DNase none detected
EEO (Electroendosmosis) ≤0.06
Endonuclease/ligase inhibitory factors none
Gel Strength ≥2000 g/cm²
Gelling Range 37 – 41 °C
Melting Range 93 – 96 °C
Moisture ≤8.5 %
Protease none detected
RNase none detected
Sulphate ≤0.06 %