AMREK296
K296-100GEA 396950 KRW
VWRCK296-100G
AMREK296
Nitrilotriacetic Acid (NTA) is a polyamino carboxylic acid which chelates di- and tri- metal cations. The chelating activity of NTA is similar to EDTA but NTA is biodegradable and easily removed during wastewater treatment.
Order Now
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Learn more About VWR