Sodium L(+)-ascorbate 99-101% USP
동의어:
Sodium ascorbate
, L(+)-Ascorbic acid sodium salt
, L-Ascorbic acid sodium salt
, Acidum ascorbicum sodium salt
0561-100GEA
204270
KRW
0561-100G
Sodium L(+)-ascorbate 99-101% USP
Sodium L(+)-ascorbate
Product is tested to USP Specifications.
포뮬러:
C₆H₇NaO₆ MW: 198,11 g/mol 녹는점: 219 °C 밀도: 1,799 g/cm³ (20 °C) 보관 온도: Ambient |
MDL 번호:
MFCD00082340 CAS 번호: 134-03-2 EINECS: 205-126-1 UN: 0000 |
사양 테스트 결과
Heavy Metals | ≤0.002% |
Loss on Drying | ≤0.25% |
pH (10%, Water) @25 °C | 7 – 8 |
Purity | 99 – 101% |
Specific Rotation | 103 – 108° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...