Methyl-4-hydroxybenzoate NF
|
Warning
|
동의어:
4-Hydroxybenzoic acid methyl ester
0572-500GEA
419880
KRW
0572-500G
Methyl-4-hydroxybenzoate NF
Methyl-4-hydroxybenzoate
Fine white crystalline powder.
포뮬러:
HOC₆H₄CO₂CH₃ MW: 152,15 g/mol 끊는점: 270 – 280 °C (1013 hPa) 녹는점: 125…128 °C 밀도: 1,1208 g/cm³ (137 °C) 보관 온도: Ambient |
MDL 번호:
MFCD00002352 CAS 번호: 99-76-3 EINECS: 202-785-7 UN: 0000 Merck 색인: 12,06182 |
사양 테스트 결과
Acidity test | Pass |
Color of solution | Pass |
Purity | 98.0 – 102.0% |
Related substances test | Pass |
Residue on ignition | ≤0.1 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...