Coliwasa trumpet, Wheaton®
공급업체: DWK Life Sciences
WHEA885235EA 436510 KRW
WHEA885235
Coliwasa trumpet, Wheaton®
샘플러 Disposable Samplers
This sampler is designed for the extraction of sludge-type or semi-solid materials.
Suitable for boring through materials.