Disinfectant spray, Pursept® A Xpress
공급업체: Brand
BRND44835EA 0 KRW
BRND44830 BRND44835
Disinfectant spray, Pursept® A Xpress
세정제 Disinfectants
For cleaning and disinfecting surfaces and equipment.
  • Manual pump-atomiser head for the 1 litre bottle is available as an accessory
  • Effective in only 15 seconds
  • Pleasant, fresh fragrance
  • Free of aldehydes
  • DGHM/VAH tested and RKI compliant, with aerosol-free application ( use with disinfectant wipes)
증명: * As recommended by the RKI, Bundesgesundheitsblatt 01/2004.