Tissue grinders, Dounce type
공급업체: DWK Life Sciences
Dura-Grind®
432-5005EA 1157300 KRW
432-5005 432-5006 432-5007
Tissue grinders, Dounce type
균질기 Tissue Grinders
Wheaton Dura-Grind® stainless steel Dounce tissue grinders provide a rugged alternative to glass Dounce tissue grinders. Smooth pestle action requires less effort than glass tissue grinders. Precision machined  No. 316 stainless steel to low tolerance of 0,0124 mm.
  • Mortar has flat bottom to stand upright
  • Single pestle and mortar are supplied as a matched set
  • Not autoclavable
Weight: 0,9 kg