GC injection port septa
VWRI548-0184EA 36460 KRW
VWRI548-0184
GC injection port septa
Long life GC septa for inlet temperature of up to 340 °C.
  • Low bleeding
  • Easy exchange, no sticking to hot surfaces
  • Good penetration and sealing properties