Phenol red : ammonium sulphate (0.002 : 0.005) % in aqueous solution Reag. Ph. Eur. 1063603, buffered pH 4.7
공급업체: VWR Chemicals
87889.260EA 243680 KRW
87889.260
Phenol red : ammonium sulphate (0.002 : 0.005) % in aqueous solution Reag. Ph. Eur. 1063603, buffered pH 4.7
Phenol red : ammonium sulphate
주문 정보:Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.
포뮬러: C₁₉H₁₃NaO₅S
보관 온도: Ambient
MDL 번호: MFCD00066901
EINECS: 252-057-8
UN: 0000