Azinphosmethyl, Extra Pure
공급업체: VWR Chemicals
Danger
123562LEA 184510 KRW
123562L
Azinphosmethyl, Extra Pure
Azinphosmethyl
포뮬러: C₁₀H₁₂N₃O₃PS₂
MW: 317,33 g/mol
끊는점: 401 °C (1013 hPa)
녹는점: 73 °C
밀도: 1,44 g/cm³ (20 °C)
CAS 번호: 86-50-0
EINECS: 201-676-1
UN: 2811
ADR: 6.1,II
Merck 색인: 12,00944