Bromothymol blue sodium salt ACS

동의어: Sodium α-(3-bromo-5-isopropyl-4-oxo-2-methyl-2,5-cyclohexadienylidene)-2-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)toluenesulphonateBromothymol blue water soluble

0554-25GEA 638170 KRW
0554-25G
Bromothymol blue sodium salt ACS
Bromothymol blue sodium salt
포뮬러: C₂₇H₂₇Br₂NaO₅S
MW: 646,37 g/mol
녹는점: 200 °C
보관 온도: Ambient
MDL 번호: MFCD00077263
CAS 번호: 34722-90-2
EINECS: 252-169-7
UN: 0000

Order Now

사양 테스트 결과

Clarity of Solution Pass
Visual Transition Interval (pH 6.0 - 7.6) Pass

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR