Cholesterol, Ultrapure
동의어:
Cholest-5-en-3-ol
0433-250GEA
1043410
KRW
0433-250G
Cholesterol, Ultrapure
Cholesterol
포뮬러:
C₂₇H₄₆O MW: 386,66 g/mol 끊는점: 360 °C 녹는점: 147 °C…149 °C 밀도: 1,07 g/cm³ (20 °C) 보관 온도: Ambient |
MDL 번호:
MFCD00003646 CAS 번호: 57-88-5 EINECS: 200-353-2 UN: 0000 Merck 색인: 13,02221 |
사양 테스트 결과
Loss on drying | ≤0.3% |
Melting point | 146−150 °C |
Residue on ignition | ≤0.1% |
Specific rotation | −38 to −34° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...