Diethylene glycol monoethyl ether, J.T.Baker®

공급업체: Avantor

동의어: 2-(2-Ethoxyethoxy)ethanolDi(ethylene glycol) ethyl ether

L216-07EA 0 KRW
L216-07
Diethylene glycol monoethyl ether, J.T.Baker®
Diethylene glycol monoethyl ether
포뮬러: C₆H₁₄O₃
MW: 134,18 g/mol
끊는점: 197 °C (1013 hPa)
녹는점: –80 °C
밀도: 0,99 g/cm³ (20 °C)
인화점: 93 °C
보관 온도: Ambient
MDL 번호: MFCD00002872
CAS 번호: 111-90-0
EINECS: 203-919-7
UN: 0000
REACH: 01-2119475105-42
Merck 색인: 13,01809

Order Now

사양 테스트 결과

For Laboratory,Research,or Manufacturing Use
Water (H₂O)(by Karl Fischer titrn) ≤0.2 %
Identification (by IR) Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR