EDTA disodium salt concentrate for dilution to 0.1 M, DILUT-IT® volumetric solution, J.T.Baker®
공급업체: Avantor
4653-01EA 1392000 KRW
4653-01 4653.
EDTA disodium salt concentrate for dilution to 0.1 M, DILUT-IT® volumetric solution, J.T.Baker®
EDTA disodium salt
(1/10 mole=33.62 g C₁₀H₁₄N₂Na₂O₈)
포뮬러: C₁₀H₁₄N₂Na₂O₈
MW: 336,21 g/mol
보관 온도: Ambient
MDL 번호: MFCD00070672
CAS 번호: 139-33-3
EINECS: 205-358-3
Merck 색인: 12,03556
Order Now
사양 테스트 결과
For Laboratory,Research,or Manufacturing Use
Molarity (by titrimetry) Passes Test
(Makes 0.1M solution after dilution to 1000 mL)