Porcine serum
공급업체: Equitech-Bio
EQUBSP30-0100HIEA 219000 KRW
EQUBSP30-0100HI EQUBSP30-0100 EQUBPS05-0100
Porcine serum
혈청
Collected from animals aged six months or older.
  • Sterility tested
  • Endotoxin and mycoplasma testing available
  • Lot to lot consistency
Raw serum is non-sterile. Low hemoglobin levels. Physiochemical constituents measured to ensure lot-to-lot consistency. Serum from specific breeds are available by request. Sterile-filtered serum is sterility tested and sterile-filtered to 0.2 micron absolute. Endotoxin and mycoplasma testing available. Heat-inactivation is available if desired.
증명: Certified U.S. origin.
Order Now
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Learn more About VWR