VWR®, Plate Tube Rotator
공급업체: VWR
VWR®
VWRA444-1173EA 0 KRW
VWRA444-1173 VWRA444-1172
VWR®, Plate Tube Rotator
쉐이커 Rotating/Rocking Shakers
VWR® Plate Tube Rotator reduces the need for frequent maintenance based on an AC synchronous motor and stably provides the required power. It has all the features and high-performance components in a compact design, allowing you to save limited benchtop space.
  • AC synchronous motor for noiseless operation
  • Accommodates up to 22 blood collection tubes, with removable plate
  • Circular rotation motion for gentle yet effective mixing
  • 30 RPM fixed speed with ±2 RPM accuracy
  • Continuous/infinite operation modes
  • Robust construction, strong metal design
  • Compact design for bench space efficiency
증명: IP protection class according to DIN EN 60529: IP 21
주문 정보:Supplied without accessories.