VWR® Mini Tube Rotator
공급업체: VWR, part of Avantor
VWRA444-1187EA 0 KRW
VWRA444-1187
VWR® Mini Tube Rotator
쉐이커 Rotating/Rocking Shakers
VWR® Mini Tube Rotator is equipped with a high-performance AC motor for quiet and efficient experiments along with a compact design.
  • Compact design for bench space efficiency
  • AC synchronous motor for noiseless operation
  • Can accommodate up to 10 Blood collection tubes
  • Gentle yet effective mixing with circular motion
  • 30 RPM fixed speed with ± 2 RPM speed accuracy
  • Continuous/Infinite operation modes
  • Robust construction, strong metal design
  • Maximum operational weight capacity of 1Kg
증명: IP Protection Class according to DIN EN 60529: IP 21
주문 정보:Supplied without accessories.
Order Now
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Learn more About VWR