J.T.Baker® BAKERBOND®, Ion Exchange Chromatography Resin, PolyPEI Multimode
공급업체: Avantor
J.T.Baker® BAKERBOND®
7585-03EA 0 KRW
7585-03 7585-02 7585-05 7585-04 7585-01
J.T.Baker® BAKERBOND®, Ion Exchange Chromatography Resin, PolyPEI Multimode
크로마토그래피 레진 HPLC Sorbents
The J.T.Baker® BAKERBOND® PolyPEI multimode ion exchange (IEX) chromatography resin functions as a weak anion exchange resin with improved selectivity for superior separation in process chromatography applications.
Powered by a proprietary surface chemistry combining weak anion exchange functionality with hydrophilicity, BAKERBOND® PolyPEI multimode cation exchange chromatography media utilizes its unique selectivity to enable higher purification performance across a wide range of biopharmaceuticals.
Designed and manufactured by Avantor to the high standards established for our J.T.Baker® brand chemicals, BAKERBOND® PolyPEI ion exchange resin can be used with other J.T.Baker® BAKERBOND® resins to enhance the separation power of your chromatography process steps.
주문 정보:1 ml and 5 mllab columns are also available under complementary products.