Methanol : Water 70:30, BAKER ANALYZED®, J.T.Baker®

Danger


Management of Change (MOC) category = N

9147-03EA 0 KRW
9147-03
Methanol : Water 70:30, BAKER ANALYZED®, J.T.Baker®
Methanol (15 - < 30%)
포뮬러: CH₄O/H₂O
보관 온도: ambient
MDL 번호: MFCD00004595
EINECS: 200-659-6
UN: 1230
ADR: 3,II

Order Now

사양 테스트 결과

For Laboratory,Research,or Manufacturing Use
Assay (CH₃OH) (v/v) 68 - 72 %
Fluorescence Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR