Penicillin G sodium salt USP

0242-1BUEA 710470 KRW
0242-1BU MOBI0242100X106UAM
Penicillin G sodium salt USP
Penicillin G sodium salt
포뮬러: C₁₆H₁₇N₂NaO₄S
MW: 356,38 g/mol
녹는점: 255 °C
밀도: 1,38 g/cm³ (20 °C)
보관 온도: Ambient
MDL 번호: MFCD00069666
CAS 번호: 69-57-8
EINECS: 200-710-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR