Lead(II) acetate basic in aqueous solution, TECHNICAL

Danger

동의어: Lead(II) hydroxide acetateAcetic acid lead (II) salt basicLead subacetate

26489.297EA 46470 KRW
26489.297
Lead(II) acetate basic in aqueous solution, TECHNICAL
Lead(II) acetate basic
포뮬러: (CH₃COO)₂Pb·2Pb(OH)₂
MW: 807,72 g/mol
밀도: 1,21 g/cm³ (20 °C)
보관 온도: Ambient
MDL 번호: MFCD00011154
CAS 번호: 1335-32-6
EINECS: 215-630-3
UN: 3082
ADR: 9,III

Order Now

사양 테스트 결과

Assay (calculated as Pb) 15 - 17 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR