Phenol red 0,4 g/L in ethanol, TECHNICAL
|
Danger
|
34304.237EA
434510
KRW
34304.237_T
34304.237
34304.237DP
Phenol red 0,4 g/L in ethanol, TECHNICAL
Phenol red
- Transition range: pH 6,4 - 8,2
포뮬러:
C₁₉H₁₄O₅S MW: 354,38 g/mol 녹는점: Min. 300 °C 보관 온도: Ambient |
MDL 번호:
MFCD00003552 CAS 번호: 143-74-8 EINECS: 205-609-7 UN: 1170 ADR: 3,II Merck 색인: 13,07329 |
사양 테스트 결과
Identification | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...