Sodium perborate tetrahydrate ≥97%, GPR RECTAPUR®

Danger

27987.295EA 76510 KRW
27987.295
Sodium perborate tetrahydrate ≥97%, GPR RECTAPUR®
Sodium perborate tetrahydrate
포뮬러: NaH₂BO₄·4H₂O
MW: 171,87 g/mol
녹는점: 60 °C
밀도: 1,73 g/cm³ (25 °C)
보관 온도: Ambient
MDL 번호: MFCD00149231
CAS 번호: 13517-20-9
EINECS: 239-172-9
UN: 0000

Order Now

사양 테스트 결과

Assay Min. 97 %
SO4 (Sulphate) Max. 0.1 %
Fe (Iron) Max. 10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR