Clarifying reagent of biological media

공급업체: VWR Chemicals
Danger

27357.232EA 299000 KRW
27357.232
Clarifying reagent of biological media
Clarifying reagent for biological media

This product has a specific odour, it may be colourless to light yellow. The yellowing does not affect adversely the clarifying efficiency of the reagent.

인화점: 20 °C (closed cup)
EINECS: 000-000-0
UN: 1993
ADR: 3,II

Order Now

사양 테스트 결과

Identification Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR