Copolymer CV/A 85/15 (copolymer vinyl chloride about 86% and vinyl acetate about 14%), TECHNICAL
27337.290EA 273030 KRW
27337.290
Copolymer CV/A 85/15 (copolymer vinyl chloride about 86% and vinyl acetate about 14%), TECHNICAL
Copolymer CV/A 85/15 (copolymer vinyl chloride about 86% and vinyl acetate about 14%)
Copolymer vinyl chloride about 86% and vinyl acetate about 14%
copolymer vinyl chloride about 86% and vinyl acetate about 14%
EINECS: 000-000-0
UN: 0000
Order Now
사양 테스트 결과
Apparent density after settling 0.3 to 0.6
Particle size Max. 4 mm