Silicone oil, Technical Grade for oil baths, VWR®
공급업체: VWR Chemicals
VWR®
24610.363EA 627130 KRW
24610.363
Silicone oil, Technical Grade for oil baths, VWR®
Silicone oil
Usable between 100 and 200 °C.
  • Essentially non-toxic
  • Environmentally safe
  • Non-halogenated
  • High thermal stability and oxidation resistance
Typical values (without guarantee)
• Appearance: crystal clear liquid
• Density (25°C): 0.96 kg/dm3
• Water content: about 30 ppm
• Viscosity (25 °C): 50 mm2/s
• Flash point (closed cup): 260 °C
• Fire point (open cup): 370 °C
보관 온도: ambient
MDL 번호: MFCD00132673
CAS 번호: 68083-14-7
EINECS: 000-000-0
UN: 0000