Iron(II) oxalate dihydrate ≥99%, purified
|
Warning
|
동의어:
Iron (II) ethanedioate dihydrate
, Oxalic acid iron (II) salt dihydrate
, Iron(II) oxalate hydrate
24179.293EA
189670
KRW
24179.293
Iron(II) oxalate dihydrate ≥99%, purified
Iron(II) oxalate dihydrate
포뮬러:
Fe(C₂O₄)·2H₂O MW: 179,9 g/mol 녹는점: 190 °C 밀도: 2,28 g/cm³ (25 °C) 보관 온도: Ambient |
MDL 번호:
MFCD00150040 CAS 번호: 6047-25-2 EINECS: 208-217-4 UN: 3288 ADR: 6.1,III |
사양 테스트 결과
Assay | Min. 99 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...