Solution for anionic surfactant determination (formerly known as Dimidium bromide-disulphine blue indicator stock solution)
공급업체: VWR Chemicals
191892HEA 409270 KRW
191892H
Solution for anionic surfactant determination (formerly known as Dimidium bromide-disulphine blue indicator stock solution)
Solution for anionic surfactant determination (formerly known as Dimidium bromide-disulphine blue indicator stock solution)
For use in the two-phase method for anionic surfactants.
Prepare the working solution as follows:
Dilute 20 ml of stock solution with 200 ml distilled water and add 20 ml of 2.5 M sulphuric acid. Dilute to 500 ml with distilled water.
주의: Store tightly closed in a cool dry place.
EINECS: 000-000-0
UN: 0000
Order Now
사양 테스트 결과
Suitability as indicator Passes test