Proclean abacus, J.T. Baker®
BAKR3432.5000EA 0 KRW
BAKR3432.5000
Proclean abacus, J.T. Baker®
Osmolality (mOsm/kg): 395 - 415
Performance Test: passes test
pH at 25°C: 8.5 - 9.1
Total Bacterial Growth (CFU/100ml): max. 5
주의: pH is measured directly after production. In course of time the actual value will decrease. This will not influence the performance of the reagent.