CyMet KX for hematology CN-free, J.T.Baker®
공급업체: Avantor
3425.0500EA
0
KRW
3425.0500
3425-00
CyMet KX for hematology CN-free, J.T.Baker®
CyMet
Lysing reagent for hematology
주의: In vitro diagnostic medical devices only for professional use.
사양 테스트 결과
Conductivity (mS) | 8.2 - 9.2 |
Osmolality (mOsm/kg) | 129 - 139 |
Performance Test | passes test |
pH at 25°C | 4.4 - 4.6 |
*) All parameters except mid cells: Test/Ref ratio > 95% and < 105%. Correlation coefficient: > 0.95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...