CyMet APR BASO II for hematology, J.T.Baker®
3479.1000PEEA
0
KRW
3479.1000PE
CyMet APR BASO II for hematology, J.T.Baker®
CyMet
Lysing reagent for hematology
주의: In vitro diagnostic medical devices only for professional use.
사양 테스트 결과
Conductivity (mS) | 12.70 - 13.70 |
Osmolality (mOsm/kg) | 227 - 257 |
Performance Test | passes test |
pH at 25°C | 3.15 - 3.25 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...