CyMet APR BASO II for hematology, J.T.Baker®
3479.1000PEEA 0 KRW
3479.1000PE
CyMet APR BASO II for hematology, J.T.Baker®
CyMet
Lysing reagent for hematology
주의: In vitro diagnostic medical devices only for professional use.
IVDLogo
Order Now
사양 테스트 결과
Conductivity (mS) 12.70 - 13.70
Osmolality (mOsm/kg) 227 - 257
Performance Test passes test
pH at 25°C 3.15 - 3.25