CyMet NR III for hematology CN-free, J.T.Baker®
공급업체: Avantor
3486.1000PEEA
0
KRW
3486-00
3486.1000PE
CyMet NR III for hematology CN-free, J.T.Baker®
CyMet
Lysing reagent for hematology
주의: In vitro diagnostic medical devices only for professional use.
사양 테스트 결과
Conductivity (mS) | 23.8 - 24.8 |
Osmolality | 307 - 317 mOsm/kg |
Performance Test | passes test |
pH at 25°C | 6.25 - 6.45 |