1-Octanesulphonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC

Warning

동의어: Octane-1-sulphonic acid sodium saltSodium 1-octansulphonateSodium 1-octanesulphonate

BDH152803UEA 424570 KRW
BDH152803U
1-Octanesulphonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC
1-Octanesulphonic acid sodium salt
포뮬러: C₈H₁₇NaO₃S
MW: 216,28 g/mol
녹는점: > 300 °C
보관 온도: Ambient
MDL 번호: MFCD00007544
CAS 번호: 5324-84-5
EINECS: 226-195-4
UN: 0000
https://kr.vwr.com/stibo/web/std.lang.all/09/50/2300950.jpg

Order Now

사양 테스트 결과

Assay (calculated on dried substance) ≥ 98,0 %
Appearance White powder
IR Spectrum Passes test
Water ≤ 2,0 %
Absorbance (200 nm) (0.25 mol/l) ≤ 0,1
Absorbance (210 nm) (0.25 mol/l) ≤ 0,05
Absorbance (220 nm) (0.25 mol/l) ≤ 0,03
Absorbance (230 nm) (0.25 mol/l) ≤ 0,02
Absorbance (240 nm) (0.25 mol/l) ≤ 0,01
Absorbance (250 nm) (0.25 mol/l) ≤ 0,01
Absorbance (260 nm) (0.25 mol/l) ≤ 0,01

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR