1-Hexanesulphonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC

동의어: Sodium hexanesulphonateSodium 1-hexanesulphonate

BDH152793MEA 425510 KRW
BDH152793M
1-Hexanesulphonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC
1-Hexanesulphonic acid sodium salt
포뮬러: C₆H₁₃O₃SNa
MW: 188,22 g/mol
녹는점: 300 °C
보관 온도: Ambient
MDL 번호: MFCD00007542
CAS 번호: 2832-45-3
EINECS: 220-601-3
UN: 0000
https://kr.vwr.com/stibo/web/std.lang.all/07/94/2300794.jpg

Order Now

사양 테스트 결과

Assay (calculated on dried substance) ≥ 99,0 %
Appearance White powder
IR Spectrum Passes test
Correlation >90% Passes test
Check best hit product IR spectrum Passes test
Water ≤ 2,0 %
Absorbance (200 nm) (0.25 mol/l) ≤ 0,1 WOU
Absorbance (210 nm) (0.25 mol/l) ≤ 0,05 WOU
Absorbance (220 nm) (0.25 mol/l) ≤ 0,04 WOU
Absorbance (230 nm) (0.25 mol/l) ≤ 0,03 WOU (K- GS)
Absorbance (240 nm) (0.25 mol/l) ≤ 0,01 WOU
Absorbance (250 nm) (0.25 mol/l) ≤ 0,01 WOU
Absorbance (260 nm) (0.25 mol/l) ≤ 0,01 WOU

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR