Eosin Y (yellowish) solution in denatured ethanol for histology, J.T.Baker®
공급업체: Avantor
Warning
동의어: Eosin yellowishTetrabromofluorescein disodium saltEosin Y disodium saltEosin 225Solvent Red 43Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoate2,4,5,7-Tetrabromofluorescein disodium saltField's Stain B
3800.1000PEEA 0 KRW
3800.1000PE 3800.2500PE
Eosin Y (yellowish) solution in denatured ethanol for histology, J.T.Baker®
Eosin Y (yellowish)
포뮬러: C₂₀H₆Br₄Na₂O₅
MW: 691,86 g/mol
녹는점: 295 to 296 °C
보관 온도: ambient
MDL 번호: MFCD00005040
CAS 번호: 17372-87-1
EINECS: 241-409-6
UN: 0000
Merck 색인: 13,03634
IVDLogo