Acrylamide ≥99.9%, Ultra Pure Grade

공급업체: VWR Collection
Danger

VWR®
VWRA0341-1KGEA 0 KRW
VWRA0341-1KG
Acrylamide ≥99.9%, Ultra Pure Grade
Acrylamide
포뮬러: C₃H₅NO
MW: 71,08 g/mol
끊는점: 232 °C (1013 hPa)
녹는점: 84,..86 °C
인화점: 138 °C
보관 온도: Ambient
MDL 번호: MFCD00008032
CAS 번호: 79-06-1
EINECS: 201-173-7
UN: 2074
ADR: 6.1,III
https://kr.vwr.com/stibo/web/std.lang.all/13/21/2301321.jpg

Order Now

사양 테스트 결과

Conductivity (40%, Water) Max. 5
DNase None
Free Acrylic acid Max. 0.001%
Identification PASS
Insolubles Max. 0.005%
Iron Max. 0.0001%
Lead Max. 0.0001%
Magnesium Max. 0.0001%
Moisture (KF) Max. 0.5%
pH (10%,0.1 M NaCl) at 25 °C 5.5 - 6.5
Protease None
Purity Min. 99.9%
RNase None

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR