2-Methoxy-1-methylethyl acetate (PGMEA) ≥99.0% (by GC) (BTS-220), J.T.Baker®
|
Danger
|
동의어:
1-Methoxy-2-propyl acetate
, DOWANOL PMA
, 2-(1-Methoxy)propyl acetate
, Propylene glycol 1-methyl ether 2-acetat
, Propylene glycol methyl ether acetat
, 1,2-Propanediol monomethyl ether acetate
6343-05EA
0
KRW
6343-05
2-Methoxy-1-methylethyl acetate (PGMEA) ≥99.0% (by GC) (BTS-220), J.T.Baker®
2-Methoxy-1-methylethyl acetate (PGMEA)
Edge bead remover (PGMEA).
포뮬러:
C₆H₁₂O₃ MW: 132,16 g/mol 끊는점: 145,8 °C (1013 hPa) 녹는점: –67 °C 밀도: 0,968 g/cm³ (20 °C) 인화점: 45 °C |
MDL 번호:
MFCD00038500 CAS 번호: 108-65-6 EINECS: 203-603-9 UN: 3272 ADR: 3,III |
사양 테스트 결과
Acidity (as CH₃COOH) | Max.0.02% |
Appearance | Pass test |
Calcium (Ca) | Max. 20 ppb |
Colour (APHA) | Max. 10 |
Iron (Fe) | Max. 20 ppb |
Magnesium (Mg) | Max. 20 ppb |
Potassium (K) | Max. 20 ppb |
Sodium (Na) | Max. 20 ppb |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...