D-(+)-Sucrose NF, Multi-Compendial low in endotoxins, J.T.Baker®

동의어: SucroseD(+)-Saccharose

4005-05EA 0 KRW
4005-05
D-(+)-Sucrose NF, Multi-Compendial low in endotoxins, J.T.Baker®
D-(+)-Sucrose

Low endotoxin. Beet derived. GMP manufactured product. The infrared spectrum conforms to the standard.

포뮬러: C₁₂H₂₂O₁₁
MW: 342,3 g/mol
녹는점: 169…170 °C
밀도: 1,29 g/cm³ (20 °C)
보관 온도: Ambient
MDL 번호: MFCD00006626
CAS 번호: 57-50-1
EINECS: 200-334-9
UN: 0000
Merck 색인: 12,09051
https://kr.vwr.com/stibo/web/std.lang.all/95/86/2299586.jpg

Order Now

사양 테스트 결과

NF - Specific Rotation [Å]²⁵ ^D (+) ≥65.9°
NF - Residue on Ignition ≤0.05%
NF - Chloride (Cl) ≤0.0035%
NF - Sulphate (SO₄) ≤0.006%
NF - Appearance (White to Off White Powder) Passes Test
NF - Calcium (Ca) Passes Test
NF - Heavy Metals (as Pb) ≤5 ppm
NF - Invert Sugar, mg ≤112
Residual Ethanol ≤5000 ppm
Residual Methanol ≤3000 ppm
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Conductivity, µS cm-1 ≤35
EP/BP - Dextrins Passes Test
EP/BP - Optical Rotation (+) 66.3 - 67.0°
EP/BP - Colour ≤45
EP/BP - Reducing Sugars Passes Test
EP/BP - Sulfite (SO₂) ≤10 ppm
EP/BP - Loss on Drying at 105 °C ≤0.1%
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - Clarity and Colour of Solution Passes Test
JP - Dextrins Passes Test
JP - Acidity or Alkalinity Passes Test
JP - Conductivity, µS cm-1 ≤35
JP - Optical Rotation (+) 66.3 - 67.0°
JP - Invert Sugar ≤0.04%
JP - Sulfite (SO₂) ≤15 ppm
JP - Lead (Pb) ≤0.5 ppm
JP - Loss on Drying at 105 °C ≤0.1%
Endotoxin Concentration (EU/g) ≤0.6
Microbial Limits (cfu/g) ≤100
Bulk Pharmaceutical Chemical
CAUTION: For Manufacturing, Processing or Repackaging
Meets BP Chemical Specifications
Meets EP Chemical Specifications
Meets JP Chemical Specifications
Meets NF Requirements
GMP Manufactured Product
Only class 2 (methanol) and class 3 (ethanol) residual solvents are present. Both are tested, quantified, and reported.
Manufactured by Ferro Pfanstiehl Laboratories for exclusive distribution by Avantor Performance Materials, Inc.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR