Methylene blue trihydrate, BAKER ANALYZED®, Certified Stain for histology, for cytology, J.T.Baker®

Warning

동의어: C.I.52015Swiss BlueBasic blue 9Methylthioninium chloride trihydrate3,7-Bis(dimethylamino)phenothiazin-5-ium chloride trihydrateMethylene blue

Q475-03EA 0 KRW
Q475-03
Methylene blue trihydrate, BAKER ANALYZED®, Certified Stain for histology, for cytology, J.T.Baker®
Methylene blue trihydrate

(C.I. 52015)
Certified for use in Histology, Bacteriology and compounding of blood stains.

포뮬러: C₁₆H₁₈ClN₃S·3H₂O
MW: 373,9 g/mol
보관 온도: Ambient
MDL 번호: MFCD00150008
CAS 번호: 7220-79-3
EINECS: 200-515-2

Order Now

사양 테스트 결과

For Laboratory,Research,or Manufacturing Use
Total Dye Content, For Information Only
Absorbance Maximum, nm, For Information Only
Absorbance at Maximum (0.6 mg/200 mL in H₂O, 1-cm path), For Information Only
Biological Test Passes Test
Certified by the Biological Stain Commission

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR