Schiff reagent (Feulgen stain form. RA Lamb), Gurr™ for microscopical staining
351204LEA
94390
KRW
351204L
Schiff reagent (Feulgen stain form. RA Lamb), Gurr™ for microscopical staining
Schiff reagent
사양 테스트 결과
Identity (colour reaction) | Passes test |
Suitability for microscopy | Passes test |
Nuclei | Blue |
Polysaccharides | Purple |
Glycogen | Purple |
Neutral mucopolysaccharides | Purple |
Mucoproteins | Purple |
Glycoproteins | Purple |
Glycolipids | Purple |
Phospholipids | Purple |
Basal membrane | Purple |
Collagen | Purple |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...