Malachite green oxalate for microscopy (C.I. 42000)
공급업체: VWR Chemicals
Danger
동의어: Bis (N-{4-[[4-(dimethylamino)phenyl](phenyl)methylene]cyclohexa-2,5-dien-1-ylidene}-N-methylmethanaminium) trioxalateBasic Green 4N,N,N',N'-Tetramethyl-4,4'-diaminotriphenylcarbenium oxalate
3076.0100EA 137450 KRW
3076.0100
Malachite green oxalate for microscopy (C.I. 42000)
Malachite green oxalate
포뮬러: C₅₂H₅₄N₄O₁₂
MW: 927,02 g/mol
녹는점: ~ 159 °C
보관 온도: ambient
MDL 번호: MFCD00011766
CAS 번호: 2437-29-8
EINECS: 219-441-7
Merck 색인: 13,05722
https://kr.vwr.com/stibo/web/std.lang.all/75/29/2297529.jpg
Order Now
사양 테스트 결과
Identification Passes test