VWR®, Bottles, Wide Neck, Clear, Round, AC Type
공급업체: VWR
VWR®
VWRU10862-360EA 80140 KRW
VWRU10862-360 VWRU10862-226 VWRU10862-358
VWR®, Bottles, Wide Neck, Clear, Round, AC Type
바틀 Glass Bottles
Clear round bottles are manufactured from soda-lime glass that conforms to USP Type III requirements.
  • No graduations
  • With black phenolic caps
Bottles may be purchased in convenience packs with closures attached to help maintain cleanliness, in bulk packs with unattached closures in separate bags, or in bulk packs without closures.