VWR®, Media/Storage Bottles, with GL Screw Caps
공급업체: VWR
VWR®
VWRU10754-826EA 137190 KRW
VWRU10754-826 VWRU10754-838 VWRU10754-816 VWRU10754-824 VWRU10754-836 VWRU10754-814 VWRU10754-828 VWRU10754-818 VWRU10754-830 VWRU10754-822 VWRU10754-820 VWRU10754-832
VWR®, Media/Storage Bottles, with GL Screw Caps
바틀 Glass Bottles
Media/Storage bottles are designed with a wide opening (30 mm internal diameter) and a drip-free polypropylene pour ring.
  • Very good chemical resistance
  • High thermal shock resistance
  • Scale graduations
  • Autoclavable
These bottles are manufactured from high quality type 1, class A, 33 expansion borosilicate glass. It can be autoclaved to 180 °C (356 °F). They feature enhanced graduations and marking spots printed in permanent white colour.
주문 정보:All bottles are available with or without a GL 45 polypropylene cap, with the exception of the 50 ml bottles, which feature a GL 32 thread.