These freestanding units are specifically designed for heavy-duty work such as pilot plant setups.
- Floor-mounted unit for heavy-duty use
- Strong but lightweight steel channel frame
- Counter-weighted cast iron feet provide sturdy support
- Feet may be bolted to the floor for added stability
Outside frame is constructed of a strong but lightweight steel channel. Interior lattice made of lab frame rods that use a horizontal channel connector; one end accommodates rod, other end fits into channel and locks tightly with a bolt.
Loosen the bolt and the connector slides easily along channel track for simple lattice adjustment. Entire apparatus is supported on sturdy, counter-weighted cast iron feet, which may be bolted to the floor for added stability in top-heavy installations.
Frame dimensions: 1219 H×1829 W mm
Overall dimensions: 1905 H×1829 W mm
주문 정보:Each lab frame includes five 1220 mm rods, three 1829 mm rods, fifteen S-connectors, sixteen channel connectors, twenty-four clamping nuts, two cast iron feet, one frame kit, and one wrench.