VWR®, Filtering Flasks, Graduated, Heavy Wall, with Glass Connector
공급업체: VWR
VWR®
VWRU10545-856EA 550430 KRW
VWRU10545-856 VWRU10545-854 VWRU10545-864 VWRU10545-852 10545-862 VWRU10545-850 VWRU10545-860 VWRU10545-858
VWR®, Filtering Flasks, Graduated, Heavy Wall, with Glass Connector
플라스크 Filter Flasks
These flasks with glass connector feature a heavy wall construction which can withstand high vacuum.
  • White enamel markings
  • Chemical resistant
Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass.
주문 정보:Tubing not included.