Mercury ≥99.6%, GPR RECTAPUR® Reag. Ph. Eur.

Danger

86136.230EA 0 KRW
86136.230
Mercury ≥99.6%, GPR RECTAPUR® Reag. Ph. Eur.
Mercury
포뮬러: Hg
MW: 200,59 g/mol
끊는점: 356.5 °C (1013 hPa)
녹는점: –39 °C
보관 온도: Ambient
MDL 번호: MFCD00011035
CAS 번호: 7439-97-6
EINECS: 231-106-7
UN: 2809
ADR: 8,III
Merck 색인: 13,05925

Order Now

사양 테스트 결과

Assay Min. 99,6 %
Insoluble in HNO₃ Max. 0,002 %
Heavy metals (as Pb) Max. 0,0005 %
Iron (Fe) Max. 0,0005 %
Loss on drying Max. 0,003 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR