Bisacrylamide ≥99.5%, Ultrapure Bioreagent for electrophoresis, purified, J.T.Baker®

Warning

동의어: N,N’-MethylenebisacrylamideN,N'-MethylenebisacrylamideBis (N,N'-methylenebisacrylamide)N,N'-MethylenediacrylamideBIS

4031-04EA 0 KRW
4031-04
Bisacrylamide ≥99.5%, Ultrapure Bioreagent for electrophoresis, purified, J.T.Baker®
Bisacrylamide

Purified for electrophoresis.

포뮬러: (H₂C=CHCONH)₂CH₂
MW: 154,17 g/mol
끊는점: 125 °C (3,33 hPa)
녹는점: 185 °C
밀도: 1,24 g/cm³
인화점: 138 °C
보관 온도: Refrigerator
MDL 번호: MFCD00008625
CAS 번호: 110-26-9
EINECS: 203-750-9
UN: 0000
https://kr.vwr.com/stibo/web/std.lang.all/70/49/2297049.jpg

Order Now

사양 테스트 결과

For Laboratory,Research,or Manufacturing Use
Assay (CH₂(NHCOCH:CH₂)₂) ≥ 99.5 %
pH of 1% Aqueous Solution at 25°C ≥ 5.3
Acrylic Acid (CH₂:CHCOOH) ≤ 0.01 %
Absorbance of a 1% Solution at 290 nm (1-cm path vs water)(au) ≤ 0.200
Conductivity of 2% Solution, µmho ≤ 10
RNase Activity Passes Test
DNase Activity (None Detected) Passes Test
Protease Activity Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR