Lead acetate cotton, StandARd®, Macron Fine Chemicals™
공급업체: Avantor Performance Materials
Danger
H220-03EA 0 KRW
H220-03
Lead acetate cotton, StandARd®, Macron Fine Chemicals™
Lead acetate cotton
In cotton balls. For arsenic.
포뮬러: (H₃CCOO)₂Pb
MW: 325,29 g/mol
녹는점: 175…180 °C
밀도: ∼2,22 g/cm³ (20 °C)
보관 온도: Ambient
CAS 번호: 6080-56-4
EINECS: 208-908-0
UN: 1616
ADR: 6.1,III
Order Now
사양 테스트 결과
For Laboratory,Research,or Manufacturing Use
Identification Passes Test