Tetramethylammonium hydroxide 25% in water, BAKER ANALYZED®, J.T. Baker®

공급업체: Avantor
Danger

V649-05EA 0 KRW
V649-05
Tetramethylammonium hydroxide 25% in water, BAKER ANALYZED®, J.T. Baker®
Tetramethylammonium hydroxide
포뮬러: C₄H₁₃NO
MW: 91,15 g/mol
끊는점: 110 °C (1013 hPa)
녹는점: 67 to 70 °C
밀도: 1,02 g/cm³ (20 °C)
보관 온도: Ambient
MDL 번호: MFCD00008280
CAS 번호: 75-59-2
EINECS: 200-882-9
UN: 1835
ADR: 8,II
Merck 색인: 14,09224

Order Now

사양 테스트 결과

For Laboratory,Research,or Manufacturing Use
Assay (C₄H₁₃NO) (by acid-base titration) 23.0 - 26.0 %
Halogens (as Cl)(meq/g) ≤ 0.01
Appearance Passes Test
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR