Methyl methacrylate ≥99% (by GC) stabilised, J.T.Baker®

공급업체: Avantor
Danger

Q690-09EA 1152000 KRW
Q690-09
Methyl methacrylate ≥99% (by GC) stabilised, J.T.Baker®
Methyl methacrylate
포뮬러: CH₂=C(CH₃)COOCH₃
MW: 100,12 g/mol
끊는점: 100 °C (1013 hPa)
녹는점: –48 °C
밀도: 0,93 g/cm³ (20 °C)
인화점: 8 °C
보관 온도: Ambient
MDL 번호: MFCD00008587
CAS 번호: 80-62-6
EINECS: 201-297-1
UN: 1247
ADR: 3,II
REACH: 01-2119452498-28
Merck 색인: 12,06005

Order Now

사양 테스트 결과

For Laboratory,Research,or Manufacturing Use
Store at or below 30 degrees C
Assay (CH₂:C(CH₃)COOCH₃) (by GC) ≥99 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR